Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QPCTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194501.2 | NP_001181430.1 | 1293 | 81 | ||
XM_015124124.2 | XP_014979610.1 | 1288 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |