Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194636.2 | NP_001181565.1 | 185 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |