Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194766.2 | NP_001181695.1 | 532 | 75 | ||
GenBank mRNA | JU320504.1 | - | 466 | 75 | ||
JU474110.1 | - | 509 | 75 | |||
JV044838.1 | - | 537 | 75 | |||
JV601370.1 | - | 519 | 75 |