Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194794.3 | NP_001181723.2 | 728 | 78 | ||
XM_015116406.2 | XP_014971892.1 | 961 | 78 | |||
GenBank mRNA | JU323404.1 | - | 738 | 78 | ||
JU473240.1 | - | 722 | 78 | |||
JV529498.1 | - | 734 | 78 | |||
JV565037.1 | - | 729 | 78 | |||
JV704655.1 | - | 731 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |