Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194847.3 | NP_001181776.1 | 648 | 75 | ||
GenBank mRNA | JV627289.1 | - | 320 | 75 | ||
JV722213.1 | - | 83 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |