Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190875.1 | NP_001177804.1 | 4045 | 63 | ||
XM_015127973.2 | XP_014983459.1 | 7050 | 63 | |||
XM_028841624.1 | XP_028697457.1 | 4514 | 63 | |||
XM_028841625.1 | XP_028697458.1 | 4685 | 63 | |||
XM_028841626.1 | XP_028697459.1 | 4380 | 63 | |||
GenBank mRNA | JV047761.1 | - | 4243 | 63 | ||
JV636286.1 | - | 4212 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |