Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194919.1 | NP_001181848.1 | 949 | 93 | ||
XM_015128460.2 | XP_014983946.1 | 1508 | 93 | |||
XM_015128461.2 | XP_014983947.1 | 1573 | 93 | |||
GenBank mRNA | JU474553.1 | - | 861 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |