Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190916.1 | NP_001177845.1 | 56 | 79 | ||
GenBank mRNA | CO774585.1 | - | 60 | 79 | ||
JU320945.1 | - | 130 | 79 | |||
JV606456.1 | - | 226 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |