Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190954.1 | NP_001177883.1 | 970 | 63 | ||
XM_015139277.2 | XP_014994763.1 | 1058 | 63 | |||
GenBank mRNA | JV053487.1 | - | 637 | 63 | ||
JV644020.1 | - | 630 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |