Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FITM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193686.1 | NP_001180615.1 | 268 | 65 | ||
XM_028827117.1 | XP_028682950.1 | 5917 | 65 | |||
XM_028827118.1 | XP_028682951.1 | 5853 | 65 | |||
XM_028827119.1 | XP_028682952.1 | 5941 | 65 | |||
XM_028827120.1 | XP_028682953.1 | 2473 | 65 | |||
GenBank mRNA | JU321899.1 | - | 247 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |