Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193696.3 | NP_001180625.2 | 472 | 59 | ||
XM_015116852.2 | XP_014972338.2 | 252 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |