Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193688.2 | NP_001180617.1 | 1143 | 104 | ||
GenBank mRNA | JU323405.1 | - | 1314 | 104 | ||
JU477487.1 | - | 1335 | 104 | |||
JV663342.1 | - | 1369 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |