Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193688.2 | NP_001180617.1 | 1603 | 85 | ||
GenBank mRNA | JU323405.1 | - | 1774 | 85 | ||
JU477487.1 | - | 1795 | 85 | |||
JV663342.1 | - | 1829 | 85 |