Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194042.1 | NP_001180971.1 | 157 | 99 | ||
XM_015127824.2 | XP_014983310.1 | 275 | 99 | |||
XM_015127826.1 | XP_014983312.1 | 73 | 99 | |||
XM_015127828.2 | XP_014983314.1 | 75 | 99 | |||
XM_028841773.1 | XP_028697606.1 | 263 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |