Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194042.1 | NP_001180971.1 | 232 | 87 | ||
XM_002806320.3 | XP_002806366.1 | 446 | 87 | |||
XM_015127822.2 | XP_014983308.1 | 458 | 87 | |||
XM_015127824.2 | XP_014983310.1 | 350 | 87 | |||
XM_015127825.2 | XP_014983311.1 | 469 | 87 | |||
XM_015127826.1 | XP_014983312.1 | 148 | 87 | |||
XM_028841772.1 | XP_028697605.1 | 457 | 87 | |||
XM_028841773.1 | XP_028697606.1 | 338 | 87 | |||
XM_028841774.1 | XP_028697607.1 | 683 | 87 | |||
GenBank mRNA | JU322459.1 | - | 455 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |