Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAIP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194156.1 | NP_001181085.1 | 331 | 109 | ||
XM_002799275.3 | XP_002799321.1 | 416 | 109 | |||
GenBank mRNA | JU319647.1 | - | 324 | 109 | ||
JU475581.1 | - | 312 | 109 | |||
JV047047.1 | - | 286 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |