Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAIP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194156.1 | NP_001181085.1 | 501 | 97 | ||
XM_002799275.3 | XP_002799321.1 | 586 | 97 | |||
GenBank mRNA | JU319647.1 | - | 494 | 97 | ||
JU475581.1 | - | 482 | 97 | |||
JV047047.1 | - | 456 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |