Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTYMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194775.2 | NP_001181704.1 | 165 | 104 | ||
XM_001094132.4 | XP_001094132.2 | 335 | 104 | |||
XM_015111561.2 | XP_014967047.1 | 168 | 104 | |||
XM_028830471.1 | XP_028686304.1 | 311 | 104 | |||
GenBank mRNA | JV053794.1 | - | 74 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |