Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194791.3 | NP_001181720.1 | 338 | 99 | ||
XM_015116205.2 | XP_014971691.1 | 597 | 99 | |||
GenBank mRNA | JV640111.1 | - | 362 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |