Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194843.1 | NP_001181772.1 | 154 | 102 | ||
GenBank mRNA | JV600761.1 | - | 126 | 102 | ||
JV668979.1 | - | 104 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |