Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194872.3 | NP_001181801.1 | 106 | 89 | ||
XM_002803907.2 | XP_002803953.1 | 285 | 89 | |||
GenBank mRNA | JU322498.1 | - | 130 | 89 | ||
JU475800.1 | - | 160 | 89 | |||
JV047229.1 | - | 196 | 89 | |||
JV668174.1 | - | 250 | 89 | |||
JV716223.1 | - | 89 |