Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHEJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245955.1 | NP_001232884.1 | 822 | 96 | ||
XM_015110986.2 | XP_014966472.1 | 969 | 96 | |||
XM_028830469.1 | XP_028686302.1 | 1154 | 96 | |||
GenBank mRNA | HM486750.1 | - | 822 | 96 | ||
JU477250.1 | - | 920 | 96 | |||
JV636380.1 | - | 920 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |