Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246671.1 | NP_001233600.1 | 1202 | 79 | ||
GenBank mRNA | GU289742.1 | - | 1202 | 79 |