Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258113.1 | NP_001245042.1 | 699 | 87 | ||
XM_015133842.2 | XP_014989328.1 | 1332 | 87 | |||
XM_015133843.2 | XP_014989329.1 | 1389 | 87 | |||
XM_015133844.2 | XP_014989330.1 | 9497 | 87 | |||
XM_015133845.2 | XP_014989331.1 | 1052 | 87 | |||
XM_015133846.1 | XP_014989332.1 | 1041 | 87 | |||
XM_015133847.1 | XP_014989333.1 | 831 | 87 | |||
XM_028844741.1 | XP_028700574.1 | 941 | 87 | |||
GenBank mRNA | JU324676.1 | - | 714 | 87 | ||
JU475328.1 | - | 699 | 87 | |||
JV046878.1 | - | 706 | 87 | |||
JV046879.1 | - | 706 | 87 |