Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258113.1 | NP_001245042.1 | 942 | 107 | ||
XM_015133842.2 | XP_014989328.1 | 1575 | 107 | |||
XM_015133843.2 | XP_014989329.1 | 1632 | 107 | |||
XM_015133844.2 | XP_014989330.1 | 9740 | 107 | |||
XM_015133845.2 | XP_014989331.1 | 1295 | 107 | |||
XM_015133846.1 | XP_014989332.1 | 1284 | 107 | |||
XM_015133847.1 | XP_014989333.1 | 1074 | 107 | |||
XM_028844741.1 | XP_028700574.1 | 1181 | 107 | |||
XM_028844742.1 | XP_028700575.1 | 795 | 107 | |||
GenBank mRNA | JU475328.1 | - | 942 | 107 | ||
JV046878.1 | - | 949 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |