Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260552.2 | NP_001247481.1 | 988 | 86 | ||
XM_015113693.2 | XP_014969179.2 | 1396 | 86 | |||
XM_015113695.2 | XP_014969181.2 | 683 | 86 | |||
XM_015113696.2 | XP_014969182.2 | 513 | 86 | |||
XM_015113697.2 | XP_014969183.2 | 482 | 86 | |||
XM_015113698.2 | XP_014969184.2 | 513 | 86 | |||
XM_015113699.2 | XP_014969185.1 | 861 | 86 | |||
XM_028832482.1 | XP_028688315.1 | 683 | 86 | |||
XM_028832483.1 | XP_028688316.1 | 482 | 86 | |||
GenBank mRNA | JU475153.1 | - | 994 | 86 | ||
JV046743.1 | - | 989 | 86 | |||
JV648189.1 | - | 1008 | 86 |