Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256898.2 | NP_001243827.2 | 499 | 65 | ||
GenBank mRNA | JU326745.1 | - | 366 | 65 | ||
JU473793.1 | - | 426 | 65 | |||
JU473794.1 | - | 426 | 65 | |||
JU473795.1 | - | 426 | 65 | |||
JV045452.1 | - | 422 | 65 | |||
JV654618.1 | - | 482 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |