Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257603.1 | NP_001244532.1 | 297 | 72 | ||
GenBank mRNA | JU320619.1 | - | 297 | 72 | ||
JU320620.1 | - | 297 | 72 | |||
JV663276.1 | - | 374 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |