Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNL4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257626.1 | NP_001244555.1 | 585 | 76 | ||
XM_015126574.2 | XP_014982060.1 | 744 | 76 | |||
GenBank mRNA | JU322032.1 | - | 585 | 76 | ||
JU472948.1 | - | 590 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |