Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257681.1 | NP_001244610.1 | 200 | 93 | ||
GenBank mRNA | JU320713.1 | - | 200 | 93 | ||
JU473738.1 | - | 239 | 93 | |||
JV043220.1 | - | 173 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |