Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFR1OP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257773.1 | NP_001244702.1 | 283 | 81 | ||
XM_015151148.2 | XP_015006634.1 | 478 | 81 | |||
GenBank mRNA | JU320035.1 | - | 283 | 81 | ||
JV045435.1 | - | 192 | 81 | |||
JV045436.1 | - | 192 | 81 | |||
JV635518.1 | - | 404 | 81 | |||
JV637318.1 | - | 404 | 81 |