Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257803.1 | NP_001244732.1 | 95 | 67 | ||
XM_015140700.2 | XP_014996186.1 | 183 | 67 | |||
GenBank mRNA | JU322593.1 | - | 95 | 67 | ||
JV638059.1 | - | 129 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |