Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM81A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257823.1 | NP_001244752.1 | 454 | 77 | ||
XM_015142337.2 | XP_014997823.1 | 633 | 77 | |||
XM_015142338.2 | XP_014997824.1 | 674 | 77 | |||
XM_015142339.2 | XP_014997825.1 | 482 | 77 | |||
XM_015142340.2 | XP_014997826.1 | 661 | 77 | |||
XM_015142341.2 | XP_014997827.1 | 702 | 77 | |||
XM_028850291.1 | XP_028706124.1 | 868 | 77 | |||
GenBank mRNA | JU333060.1 | - | 454 | 77 | ||
JV606628.1 | - | 587 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |