Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257838.1 | NP_001244767.1 | 970 | 69 | ||
GenBank mRNA | JU335376.1 | - | 970 | 69 | ||
JU476322.1 | - | 936 | 69 | |||
JV045784.1 | - | 936 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |