Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYKPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260930.2 | NP_001247859.1 | 457 | 60 | ||
XM_015141558.2 | XP_014997044.1 | 624 | 60 | |||
XM_028849166.1 | XP_028704999.1 | 612 | 60 | |||
XM_028849167.1 | XP_028705000.1 | 564 | 60 | |||
XM_028849168.1 | XP_028705001.1 | 623 | 60 | |||
XM_028849169.1 | XP_028705002.1 | 624 | 60 | |||
XM_028849170.1 | XP_028705003.1 | 333 | 60 | |||
GenBank mRNA | JU322930.1 | - | 457 | 60 | ||
JU478189.1 | - | 531 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |