Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261071.1 | NP_001248000.1 | 449 | 113 | ||
XM_015115704.2 | XP_014971190.1 | 572 | 113 | |||
XM_015115705.2 | XP_014971191.1 | 473 | 113 | |||
GenBank mRNA | JU332435.1 | - | 449 | 113 | ||
JU473900.1 | - | 449 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |