Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261697.1 | NP_001248626.1 | 316 | 72 | ||
GenBank mRNA | JU320304.1 | - | 311 | 72 | ||
JU320305.1 | - | 311 | 72 | |||
JV043513.1 | - | 316 | 72 | |||
JV637803.1 | - | 318 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |