Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261699.1 | NP_001248628.1 | 2557 | 99 | ||
GenBank mRNA | JU322325.1 | - | 2557 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |