Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261745.1 | NP_001248674.1 | 425 | 119 | ||
XM_015114513.2 | XP_014969999.1 | 463 | 119 | |||
GenBank mRNA | JU323308.1 | - | 413 | 119 | ||
JV046157.1 | - | 425 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |