Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPIND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265760.2 | NP_001252689.1 | 1330 | 72 | ||
XM_015149746.2 | XP_015005232.1 | 1589 | 72 | |||
GenBank mRNA | JV504584.1 | - | 1008 | 72 | ||
JV600210.1 | - | 1333 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |