Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265799.1 | NP_001252728.1 | 1386 | 58 | ||
GenBank mRNA | JV506362.1 | - | 257 | 58 | ||
JV667398.1 | - | 1386 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |