Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265803.1 | NP_001252732.1 | 346 | 107 | ||
GenBank mRNA | JV667338.1 | - | 346 | 107 | ||
JV706336.1 | - | 338 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |