Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265866.1 | NP_001252795.1 | 366 | 113 | ||
GenBank mRNA | JU321276.1 | - | 409 | 113 | ||
JU473663.1 | - | 416 | 113 | |||
JV045529.1 | - | 366 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |