Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265945.1 | NP_001252874.1 | 586 | 63 | ||
XM_015123954.2 | XP_014979440.2 | 917 | 63 | |||
XM_015123956.2 | XP_014979442.2 | 738 | 63 | |||
XM_028838385.1 | XP_028694218.1 | 614 | 63 | |||
XM_028838386.1 | XP_028694219.1 | 1031 | 63 | |||
GenBank mRNA | JU323098.1 | - | 578 | 63 | ||
JU471714.1 | - | 576 | 63 | |||
JV048291.1 | - | 586 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |