Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265945.1 | NP_001252874.1 | 812 | 112 | ||
GenBank mRNA | JU323098.1 | - | 804 | 112 | ||
JU471714.1 | - | 802 | 112 | |||
JV048291.1 | - | 812 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |