Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265960.1 | NP_001252889.1 | 665 | 77 | ||
GenBank mRNA | JU336223.1 | - | 651 | 77 | ||
JU477060.1 | - | 664 | 77 | |||
JV048150.1 | - | 665 | 77 | |||
JV602090.1 | - | 777 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |