Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASTOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266104.1 | NP_001253033.1 | 236 | 83 | ||
GenBank mRNA | JU328414.1 | - | 183 | 83 | ||
JU477044.1 | - | 236 | 83 | |||
JV480559.1 | - | 191 | 83 |