Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266335.1 | NP_001253264.1 | 722 | 56 | ||
XM_015115452.2 | XP_014970938.1 | 1112 | 56 | |||
GenBank mRNA | JU332642.1 | - | 703 | 56 | ||
JV046011.1 | - | 833 | 56 | |||
JV640768.1 | - | 914 | 56 | |||
JV667331.1 | - | 722 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |