Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266365.2 | NP_001253294.1 | 347 | 81 | ||
XM_015136365.2 | XP_014991851.1 | 969 | 81 | |||
XM_028846690.1 | XP_028702523.1 | 1002 | 81 | |||
XM_028846691.1 | XP_028702524.1 | 594 | 81 | |||
GenBank mRNA | JV565026.1 | - | 349 | 81 |
Gene Symbol: ABCC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV726246.1 | - | 4054 | 81 |