Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FYTTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266475.1 | NP_001253404.1 | 133 | 76 | ||
XM_028843373.1 | XP_028699206.1 | 214 | 76 | |||
XM_028843374.1 | XP_028699207.1 | 214 | 76 | |||
GenBank mRNA | JU322450.1 | - | 176 | 76 | ||
JU473399.1 | - | 176 | 76 | |||
JV045407.1 | - | 133 | 76 |